siRNA Details
Gene Symbol: COPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006837.2 | NP_006828.2 | 2 | 605 | ||
GenBank mRNA | AB209134.1 | - | 1 | 2315 | ||
AK056967.1 | - | 4 | 381 | |||
AK128519.1 | - | 1 | 2597 | |||
AK310434.1 | - | 2 | 407 | |||
AK311071.1 | - | 2 | 409 | |||
BC001187.1 | - | 2 | 409 | 3354130 | ||
BC001859.2 | - | 2 | 387 | 3530890 | ||
BC007272.1 | - | 2 | 383 | 3161025 | ||
BI752436.1 | - | 595 | ||||
BX648542.1 | - | 2 | 2942 | |||
CR541678.1 | - | 2 | 274 | |||
U65928.1 | - | 2 | 401 | |||
U70734.1 | - | 2 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
