siRNA Details
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 20 | 4842 | ||
NM_014903.5 | NP_055718.4 | 20 | 4842 | |||
XM_005269215.3 | XP_005269272.1 | 20 | 8843 | |||
XM_011538940.2 | XP_011537242.1 | 19 | 8802 | |||
XM_011538941.2 | XP_011537243.1 | 19 | 8801 | |||
XM_011538944.2 | XP_011537246.1 | 18 | 8310 | |||
XM_017020164.1 | XP_016875653.1 | 20 | 8845 | |||
XM_017020165.1 | XP_016875654.1 | 19 | 8803 | |||
XM_017020166.1 | XP_016875655.1 | 20 | 4831 | |||
XM_017020167.1 | XP_016875656.1 | 19 | 4762 | |||
XM_017020168.1 | XP_016875657.1 | 19 | 8355 | |||
XM_017020169.1 | XP_016875658.1 | 18 | 8312 | |||
XM_017020170.1 | XP_016875659.1 | 18 | 8309 | |||
XM_017020171.1 | XP_016875660.1 | 12 | 8138 | |||
XM_017020172.1 | XP_016875661.1 | 11 | 3734 | |||
XM_017020173.1 | XP_016875662.1 | 11 | 7647 | |||
XM_017020174.1 | XP_016875663.1 | 10 | 7605 | |||
XM_017020175.1 | XP_016875664.1 | 11 | 3526 | |||
GenBank mRNA | AB023155.2 | - | 5 | 1352 | ||
AF397731.1 | - | 19 | 4426 | |||
AJ488201.1 | - | 20 | 4714 | |||
AK298525.1 | - | 4 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
