siRNA Details
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 28 | 5498 | ||
XM_017020164.1 | XP_016875653.1 | 27 | 9480 | |||
XM_017020165.1 | XP_016875654.1 | 26 | 9438 | |||
XM_017020166.1 | XP_016875655.1 | 28 | 5487 | |||
XM_017020167.1 | XP_016875656.1 | 27 | 5418 | |||
XM_017020168.1 | XP_016875657.1 | 27 | 9011 | |||
XM_017020169.1 | XP_016875658.1 | 26 | 8968 | |||
XM_017020171.1 | XP_016875660.1 | 20 | 8794 | |||
XM_017020173.1 | XP_016875662.1 | 19 | 8303 | |||
XM_017020174.1 | XP_016875663.1 | 18 | 8261 | |||
GenBank mRNA | AJ488201.1 | - | 28 | 5370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
