siRNA Details
Gene Symbol: USP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282659.1 | NP_001269588.1 | 19, 20 | 3084 | ||
NM_017944.3 | NP_060414.3 | 18, 19 | 2880 | |||
XM_005252997.1 | XP_005253054.1 | 20, 21 | 3144 | |||
XM_011520196.1 | XP_011518498.1 | 12, 13 | 2259 | |||
XM_017017953.1 | XP_016873442.1 | 20, 21 | 3200 | |||
XM_017017954.1 | XP_016873443.1 | 12, 13 | 3407 | |||
GenBank mRNA | AF059318.1 | - | 2, 3 | 175 | ||
AK027362.1 | - | 18, 19 | 2285 | |||
AK074076.1 | - | 4, 5 | 572 | |||
AK092290.1 | - | 20, 21 | 2500 | |||
AK127264.1 | - | 19, 20 | 2880 | |||
AL832991.2 | - | 4, 5 | 495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
