siRNA Details
Gene Symbol: NSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022455.4 | NP_071900.2 | 5 | 2414 | ||
NM_172349.2 | NP_758859.1 | 6 | 1629 | |||
GenBank mRNA | AF322907.1 | - | 2 | 1985 | ||
AF380302.1 | - | 6 | 1570 | |||
AF395588.1 | - | 5 | 2321 | |||
AK056667.1 | - | 4 | 1393 | |||
AK091358.1 | - | 1 | 221 | |||
AY049721.1 | - | 4 | 2294 | |||
BC139789.1 | - | 5 | 1499 | |||
BC144629.1 | - | 6 | 1859 | |||
BC150628.1 | - | 5 | 1504 | 9056998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
