siRNA Details
Gene Symbol: SETD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160305.2 | NP_001153777.1 | 8 | 1288 | ||
NM_024860.3 | NP_079136.2 | 9 | 1216 | |||
NR_134583.1 | 9 | 1247 | ||||
GenBank mRNA | AK024801.1 | - | 9 | 1172 | ||
AK127136.1 | - | 5 | 2693 | |||
AK290495.1 | - | 8 | 1250 | |||
BC022451.1 | - | 8 | 1250 | 4794299 | ||
BC110433.1 | - | 9 | 1231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
