siRNA Details
Gene Symbol: USP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321291.1 | NP_001308220.1 | 4 | 698 | ||
XM_005257542.2 | XP_005257599.1 | 4 | 722 | |||
XM_005257543.2 | XP_005257600.1 | 4 | 722 | |||
XM_011525065.1 | XP_011523367.1 | 4 | 733 | |||
XM_011525066.2 | XP_011523368.1 | 4 | 595 | |||
XM_011525067.2 | XP_011523369.1 | 4 | 595 | |||
XM_011525068.1 | XP_011523370.1 | 4 | 636 | |||
XM_011525069.2 | XP_011523371.1 | 4 | 595 | |||
XM_011525070.1 | XP_011523372.1 | 4 | 595 | |||
XM_011525071.2 | XP_011523373.1 | 4 | 595 | |||
XM_017024900.1 | XP_016880389.1 | 4 | 722 | |||
XM_017024901.1 | XP_016880390.1 | 2 | 257 | |||
GenBank mRNA | AB040886.1 | - | 4 | 615 | ||
AK001671.1 | - | 4 | 733 | |||
AK022913.1 | - | 4 | 640 | |||
AK225155.1 | - | 4 | 640 | |||
AK225426.1 | - | 4 | 597 | |||
AY169386.1 | - | 2 | 397 | |||
BC016487.1 | - | 4 | 699 | |||
BC027992.1 | - | 4 | 636 | |||
BC038983.1 | - | 4 | 707 | |||
BC071582.1 | - | 4 | 698 | 30345255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
