siRNA Details
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002221.3 | NP_002212.3 | 3 | 2294 | ||
GenBank mRNA | AJ242780.1 | - | 2 | 2011 | ||
AK289700.1 | - | 2 | 740 | |||
AK313801.1 | - | 2 | 759 | |||
AY513281.1 | - | 3 | 535 | |||
BQ434930.1 | - | 2 | 446 | |||
X57206.1 | - | 2 | 633 | |||
Y18024.2 | - | 2 | 1954 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
