siRNA Details
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030648.3 | NP_085151.1 | 8 | 1968 | ||
XM_017008661.1 | XP_016864150.1 | 6 | 1061 | |||
GenBank mRNA | AB051504.1 | - | 8 | 1603 | ||
AF448510.1 | - | 8 | 1603 | |||
BC121055.1 | - | 8 | 1512 | 40118363 | ||
BC121056.1 | - | 8 | 1512 | 40118365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
