siRNA Details
Gene Symbol: UBE2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058167.2 | NP_477515.2 | 7 | 2146 | ||
NM_194315.1 | NP_919296.1 | 8 | 2194 | |||
NM_194457.1 | NP_919439.1 | 6 | 2015 | |||
NM_194458.1 | NP_919440.1 | 7 | 2275 | |||
XM_005244718.3 | XP_005244775.1 | 7 | 2424 | |||
XM_005244719.4 | XP_005244776.1 | 7 | 2312 | |||
XM_006710333.3 | XP_006710396.1 | 7 | 2591 | |||
XM_011540613.2 | XP_011538915.1 | 6 | 2019 | |||
XM_011540614.2 | XP_011538916.1 | 7 | 2377 | |||
XM_011540616.2 | XP_011538918.1 | 8 | 2386 | |||
XM_011540617.2 | XP_011538919.1 | 7 | 2153 | |||
XM_017000238.1 | XP_016855727.1 | 8 | 2257 | |||
XM_017000239.1 | XP_016855728.1 | 6 | 2475 | |||
GenBank mRNA | AF116687.2 | - | 1 | 510 | ||
AK056065.1 | - | 7 | 2055 | |||
AK098803.1 | - | 2 | 1512 | |||
BC094777.1 | - | 7 | 2145 | 30340049 | ||
BE515187.1 | - | 2 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
