siRNA Details
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323303.1 | NP_001310232.1 | 7 | 1158 | ||
NM_001323304.1 | NP_001310233.1 | 7 | 921 | |||
NM_001323305.1 | NP_001310234.1 | 8 | 1031 | |||
NM_003600.3 | NP_003591.2 | 6 | 1048 | |||
NM_198433.2 | NP_940835.1 | 8 | 1256 | |||
NM_198434.2 | NP_940836.1 | 7 | 947 | |||
NM_198435.2 | NP_940837.1 | 6 | 837 | |||
NM_198436.2 | NP_940838.1 | 7 | 933 | |||
NM_198437.2 | NP_940839.1 | 6 | 823 | |||
XM_017028034.1 | XP_016883523.1 | 8 | 1378 | |||
XM_017028035.1 | XP_016883524.1 | 6 | 738 | |||
GenBank mRNA | AF008551.1 | - | 5 | 648 | ||
AF011468.1 | - | 6 | 904 | |||
AK301769.1 | - | 6 | 510 | |||
AK312776.1 | - | 6 | 715 | |||
BC001280.1 | - | 7 | 780 | 3456017 | ||
BC002499.2 | - | 6 | 776 | 3051177 | ||
BC006423.1 | - | 6 | 673 | 3532438 | ||
BC027464.1 | - | 7 | 815 | 4449474 | ||
BQ435059.1 | - | 6 | 669 | |||
D84212.1 | - | 7 | 814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
