siRNA Details
Gene Symbol: UBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003968.3 | NP_003959.3 | 18 | 1677 | ||
NM_198195.1 | NP_937838.1 | 17 | 1635 | |||
XM_011534210.1 | XP_011532512.1 | 17 | 1591 | |||
XM_011534211.1 | XP_011532513.1 | 16 | 1544 | |||
GenBank mRNA | AF046024.1 | - | 17 | 1680 | ||
AK002159.1 | - | 17 | 1622 | |||
AL117566.1 | - | 18 | 1673 | |||
BC022853.1 | - | 18 | 1677 | 4073511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
