siRNA Details
Gene Symbol: UBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003968.3 | NP_003959.3 | 9 | 650 | ||
NM_198195.1 | NP_937838.1 | 8 | 608 | |||
XM_011534210.1 | XP_011532512.1 | 8 | 564 | |||
XM_011534211.1 | XP_011532513.1 | 7 | 517 | |||
GenBank mRNA | AB012190.1 | - | 7 | 567 | ||
AF046024.1 | - | 8 | 651 | |||
AK002159.1 | - | 8 | 595 | |||
AK289392.1 | - | 9 | 636 | |||
AK298878.1 | - | 4 | 323 | |||
AK314751.1 | - | 9 | 633 | |||
AK316540.1 | - | 6 | 487 | |||
AL117566.1 | - | 9 | 646 | |||
BC022853.1 | - | 9 | 649 | 4073511 | ||
CR533537.1 | - | 9 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
