siRNA Details
Gene Symbol: SMYD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006062.2 | NP_006053.2 | 6 | 643 | ||
XM_006711918.3 | XP_006711981.1 | 6 | 730 | |||
XM_017003163.1 | XP_016858652.1 | 5 | 2363 | |||
GenBank mRNA | AK300779.1 | - | 6 | 617 | ||
AK301809.1 | - | 6 | 636 | |||
BC073806.1 | - | 6 | 619 | 6671194 | ||
U50383.1 | - | 6 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
