siRNA Details
Gene Symbol: EHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289413.1 | NP_001276342.1 | 17, 18 | 2529 | ||
NM_006709.4 | NP_006700.3 | 19, 20 | 2486 | |||
NM_025256.6 | NP_079532.5 | 18, 19 | 2384 | |||
XM_005248824.3 | XP_005248881.1 | 18, 19 | 2627 | |||
GenBank mRNA | AJ315532.1 | - | 19, 20 | 2456 | ||
AK092866.1 | - | 2, 3 | 1401 | |||
AK302904.1 | - | 17, 18 | 2529 | |||
BC002686.1 | - | 16, 17 | 1956 | |||
BC009351.1 | - | 19, 20 | 2453 | |||
BC018718.1 | - | 19, 20 | 2444 | 4123580 | ||
BC020970.1 | - | 18, 19 | 2421 | |||
X69838.1 | - | 15, 16 | 1876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
