siRNA Details
Gene Symbol: OTUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017670.2 | NP_060140.2 | 5, 6 | 1034 | ||
NR_003089.1 | 4, 5 | 1242 | ||||
GenBank mRNA | AF161381.1 | - | 5, 6 | 442 | ||
AK000120.1 | - | 5, 6 | 474 | |||
AK098029.1 | - | 4, 5 | 1242 | |||
AK225133.1 | - | 5, 6 | 441 | |||
AK298287.1 | - | 6, 7 | 753 | |||
AK303228.1 | - | 4, 5 | 684 | |||
AK307413.1 | - | 5, 6 | 569 | |||
AL096714.1 | - | 4, 5 | 653 | |||
AY177200.1 | - | 5, 6 | 430 | |||
BC007519.2 | - | 5, 6 | 438 | 3051266 | ||
BC010368.1 | - | 5, 6 | 460 | 4334279 | ||
BC107701.1 | - | 5, 6 | 460 | 6494629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
