siRNA Details
Gene Symbol: OTUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017670.2 | NP_060140.2 | 7 | 2265 | ||
NR_003089.1 | 6 | 2473 | ||||
GenBank mRNA | AK000120.1 | - | 7 | 1705 | ||
AL096714.1 | - | 6 | 1884 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_134150, BC054410, BC022575, AK145273, AK145970.
Gene Ontology Categories:
