siRNA Details
Gene Symbol: PRDM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020228.2 | NP_064613.2 | 16 | 2490 | ||
NM_199437.1 | NP_955469.1 | 15 | 2478 | |||
NM_199438.1 | NP_955470.1 | 12 | 2216 | |||
NM_199439.1 | NP_955471.1 | 11 | 2204 | |||
XM_011542912.2 | XP_011541214.1 | 15 | 2740 | |||
XM_017018023.1 | XP_016873512.1 | 11 | 4289 | |||
GenBank mRNA | AB033057.2 | - | 12 | 2000 | ||
AF275817.1 | - | 12 | 2216 | |||
AF503171.1 | - | 11 | 2135 | |||
AK000234.1 | - | 9 | 1445 | |||
AK310354.1 | - | 16 | 2350 | |||
AL833172.1 | - | 15 | 2481 | |||
BC112934.1 | - | 15 | 2470 | 6502888 | ||
BC117415.1 | - | 12 | 2055 | 40125966 | ||
BC143612.1 | - | 12 | 2055 | 9052125 | ||
BC143613.1 | - | 11 | 2043 | 9052126 | ||
BX648944.1 | - | 16 | 2491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
