siRNA Details
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 13 | 2206 | ||
NM_005474.4 | NP_005465.2 | 13 | 2203 | |||
XM_005256905.1 | XP_005256962.1 | 13 | 2201 | |||
XM_005256906.4 | XP_005256963.1 | 13 | 2162 | |||
XM_011524149.1 | XP_011522451.1 | 13 | 2177 | |||
XM_011524150.1 | XP_011522452.1 | 12 | 1953 | |||
XM_017023988.1 | XP_016879477.1 | 15 | 2315 | |||
XM_017023989.1 | XP_016879478.1 | 15 | 2281 | |||
XM_017023990.1 | XP_016879479.1 | 15 | 2306 | |||
XM_017023991.1 | XP_016879480.1 | 15 | 2312 | |||
XM_017023992.1 | XP_016879481.1 | 15 | 2301 | |||
XM_017023993.1 | XP_016879482.1 | 15 | 2288 | |||
XM_017023994.1 | XP_016879483.1 | 11 | 2269 | |||
XM_017023995.1 | XP_016879484.1 | 13 | 2171 | |||
XM_017023996.1 | XP_016879485.1 | 14 | 2378 | |||
GenBank mRNA | AB011172.2 | - | 13 | 2175 | ||
AB177853.1 | - | 3 | 655 | |||
AF039691.1 | - | 8 | 1308 | |||
AF132608.1 | - | 12 | 1871 | |||
AK075095.1 | - | 4 | 852 | |||
AK294760.1 | - | 13 | 2171 | |||
BC038423.1 | - | 2 | 485 | |||
BC051824.1 | - | 13 | 2082 | 6043491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
