siRNA Details
Gene Symbol: COPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006837.2 | NP_006828.2 | 6 | 1056 | ||
GenBank mRNA | AB209134.1 | - | 5 | 2766 | ||
AK128519.1 | - | 5 | 3048 | |||
AK310434.1 | - | 6 | 858 | |||
AK311071.1 | - | 6 | 873 | |||
BC001187.1 | - | 6 | 860 | 3354130 | ||
BC001859.2 | - | 6 | 838 | 3530890 | ||
BC007272.1 | - | 6 | 834 | 3161025 | ||
BX648542.1 | - | 6 | 3393 | |||
CR541678.1 | - | 6 | 725 | |||
U65928.1 | - | 6 | 852 | |||
U70734.1 | - | 6 | 845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
