siRNA Details
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306199.1 | NP_001293128.1 | 4 | 1023 | ||
NM_030648.3 | NP_085151.1 | 4 | 1023 | |||
XM_017008661.1 | XP_016864150.1 | 2 | 116 | |||
GenBank mRNA | AB051504.1 | - | 4 | 658 | ||
AF448510.1 | - | 4 | 658 | |||
AF462150.1 | - | 4 | 391 | |||
AK310856.1 | - | 4 | 492 | |||
BC121055.1 | - | 4 | 567 | 40118363 | ||
BC121056.1 | - | 4 | 567 | 40118365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
