siRNA Details
Gene Symbol: NUAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030952.2 | NP_112214.2 | 6, 7 | 982 | ||
XM_005245515.4 | XP_005245572.1 | 2, 3 | 239 | |||
GenBank mRNA | AK074830.1 | - | 6, 7 | 936 | ||
AK303577.1 | - | 6, 7 | 982 | |||
AL136891.1 | - | 6, 7 | 946 | |||
BC017306.2 | - | 6, 7 | 918 | 5088037 | ||
CR749209.1 | - | 2, 3 | 4411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
