siRNA Details
Gene Symbol: SETD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032233.2 | NP_115609.2 | 2 | 242 | ||
NM_199123.1 | NP_954574.1 | 2 | 242 | |||
XM_005268127.3 | XP_005268184.1 | 2 | 244 | |||
XM_011537231.2 | XP_011535533.2 | 2 | 194 | |||
XM_011537232.2 | XP_011535534.1 | 2 | 1372 | |||
XM_011537233.2 | XP_011535535.1 | 2 | 194 | |||
XM_011537234.1 | XP_011535536.1 | 2 | 111 | |||
XM_011537235.1 | XP_011535537.1 | 2 | 194 | |||
XM_017021699.1 | XP_016877188.1 | 1 | 90 | |||
XM_017021700.1 | XP_016877189.1 | 2 | 278 | |||
XM_017021701.1 | XP_016877190.1 | 2 | 244 | |||
XM_017021702.1 | XP_016877191.1 | 2 | 212 | |||
GenBank mRNA | AK302882.1 | - | 2 | 1585 | ||
BC066931.1 | - | 2 | 143 | |||
BC120967.1 | - | 2 | 97 | |||
BC120968.2 | - | 2 | 97 | |||
BC127624.1 | - | 2 | 97 | |||
BC127625.1 | - | 2 | 97 | |||
BC142995.1 | - | 2 | 254 | 8860450 | ||
BC148251.1 | - | 1 | 93 | |||
BX161441.1 | - | 2 | 141 | |||
BX161471.1 | - | 2 | 232 | |||
CB988965.1 | - | 2 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
