siRNA Details
Gene Symbol: CLEC4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014358.3 | NP_055173.1 | 6 | 1684 | ||
GenBank mRNA | AB024718.1 | - | 6 | 1669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
