siRNA Details
Gene Symbol: PPY2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002181.1 | 1 | 127 | |||
GenBank mRNA | AF085945.1 | - | 1 | 22 | ||
AF222903.1 | - | 1 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
