siRNA Details
Gene Symbol: HTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000200.2 | NP_000191.1 | 5, 6 | 314 | ||
GenBank mRNA | BC009791.1 | - | 5, 6 | 272 | 4293405 | |
BC095438.1 | - | 5, 6 | 271 | 30352697 | ||
BP332860.1 | - | 5, 6 | 268 | |||
CB957931.1 | - | 5, 6 | 314 | |||
M18372.1 | - | 4, 5 | 234 | |||
M26665.1 | - | 5, 6 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
