siRNA Details
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002059.4 | NP_002050.1 | 5 | 701 | ||
NM_022556.3 | NP_072050.1 | 5 | 656 | |||
NM_022557.3 | NP_072051.1 | 4 | 954 | |||
NM_022558.3 | NP_072052.1 | 5 | 697 | |||
GenBank mRNA | AB451332.1 | - | 4 | 515 | ||
AB451477.1 | - | 4 | 515 | |||
AF006060.1 | - | 5 | 527 | |||
AF006061.1 | - | 4 | 556 | |||
BC020760.1 | - | 5 | 622 | 4720475 | ||
CR541902.1 | - | 4 | 560 | |||
CR541924.1 | - | 4 | 560 | |||
J03756.1 | - | 4 | 874 | |||
M38451.1 | - | 5 | 572 |
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 5 | 710 | ||
GenBank mRNA | AK291587.1 | - | 5 | 629 | ||
BC002717.2 | - | 5 | 603 | 3631916 | ||
BC005921.1 | - | 5 | 622 | 4043768 | ||
BC020756.1 | - | 5 | 678 | 4769769 | ||
BC057768.1 | - | 5 | 622 | 30337725 | ||
BC062775.1 | - | 5 | 626 | 4604848 | ||
BT006926.1 | - | 4 | 560 | |||
J00118.1 | - | 5 | 589 | |||
M25118.1 | - | 1 | 83 | |||
V00593.1 | - | 4 | 412 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 5 | 675 | ||
NM_022644.3 | NP_072170.1 | 4 | 928 | |||
NM_022645.2 | NP_072171.1 | 3 | 390 | |||
GenBank mRNA | BC022044.1 | - | 5 | 626 | 4769046 | |
BC035965.1 | - | 5 | 622 | 4731880 | ||
BC119748.2 | - | 5 | 619 | 40119242 | ||
V00573.1 | - | 5 | 584 |
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 5 | 636 | ||
NM_022559.3 | NP_072053.1 | 5 | 591 | |||
NM_022560.3 | NP_072054.1 | 4 | 516 | |||
GenBank mRNA | A00469.1 | - | 5 | 599 | ||
AF110644.1 | - | 5 | 432 | |||
AF185611.1 | - | 6 | 464 | |||
AV753220.1 | - | 3 | 456 | |||
AY613431.1 | - | 5 | 622 | |||
AY613432.1 | - | 5 | 622 | |||
BC062475.1 | - | 5 | 556 | 30393581 | ||
BC075012.2 | - | 5 | 587 | 30915404 | ||
BC075013.2 | - | 5 | 587 | 30915617 | ||
BC090045.1 | - | 5 | 603 | 30394115 | ||
CD108592.1 | - | 4 | 482 | |||
V00519.1 | - | 5 | 600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
