siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 5 | 815 | ||
NM_022559.3 | NP_072053.1 | 5 | 770 | |||
NM_022560.3 | NP_072054.1 | 4 | 695 | |||
GenBank mRNA | A00469.1 | - | 5 | 778 | ||
AF110644.1 | - | 5 | 611 | |||
AV753220.1 | - | 3 | 635 | |||
AY613431.1 | - | 5 | 801 | |||
AY613432.1 | - | 5 | 801 | |||
BC062475.1 | - | 5 | 735 | 30393581 | ||
BC090045.1 | - | 5 | 782 | 30394115 | ||
CD108592.1 | - | 4 | 661 | |||
V00519.1 | - | 5 | 779 |
Gene Symbol: ACER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092487.1 | - | 2045 | 30528463 |
Gene Symbol: ANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092527.1 | - | 2278 |
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 5 | 887 | ||
GenBank mRNA | AK291587.1 | - | 5 | 806 | ||
BC002717.2 | - | 5 | 780 | 3631916 | ||
BC005921.1 | - | 5 | 799 | 4043768 | ||
BC020756.1 | - | 5 | 855 | 4769769 | ||
BC057768.1 | - | 5 | 799 | 30337725 | ||
BC062775.1 | - | 5 | 803 | 4604848 | ||
J00118.1 | - | 5 | 767 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 5 | 857 | ||
NM_022644.3 | NP_072170.1 | 4 | 1110 | |||
NM_022645.2 | NP_072171.1 | 3 | 572 | |||
GenBank mRNA | BC022044.1 | - | 5 | 808 | 4769046 | |
BC035965.1 | - | 5 | 804 | 4731880 | ||
V00573.1 | - | 5 | 766 |
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX426214.2 | - | 5 | 680 |
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002059.4 | NP_002050.1 | 5 | 880 | ||
NM_022556.3 | NP_072050.1 | 5 | 835 | |||
NM_022557.3 | NP_072051.1 | 4 | 1133 | |||
NM_022558.3 | NP_072052.1 | 5 | 876 | |||
GenBank mRNA | AF006061.1 | - | 4 | 735 | ||
BC020760.1 | - | 5 | 801 | 4720475 | ||
J03756.1 | - | 4 | 1053 |
Gene Symbol: RPL36AP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020828.1 | - | 1241 | 4769647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
