siRNA Details
Gene Symbol: IFNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002172.2 | NP_002163.2 | 1 | 144 | ||
GenBank mRNA | BC074956.2 | - | 1 | 144 | 30915281 | |
BC104159.1 | - | 1 | 142 | 40029547 | ||
BC104160.1 | - | 1 | 142 | 40029549 | ||
V00542.1 | - | 1 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
