siRNA Details
Gene Symbol: PF4V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002620.3 | NP_002611.1 | 3 | 386 | ||
GenBank mRNA | BC130653.1 | - | 3 | 347 | 40146684 | |
BC130657.1 | - | 3 | 347 | 40146688 |
Gene Symbol: PF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002619.3 | NP_002610.1 | 3 | 481 | ||
XM_005265696.3 | XP_005265753.1 | 3 | 382 | |||
GenBank mRNA | AJ705588.1 | - | 3 | 355 | ||
BC093965.1 | - | 3 | 316 | 7939810 | ||
BC112093.1 | - | 3 | 316 | 8327561 | ||
M25897.1 | - | 3 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
