siRNA Details
Gene Symbol: CCL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032965.5 | NP_116741.2 | 2 | 638 | ||
GenBank mRNA | AF031587.1 | - | 2 | 92 | ||
BX098368.1 | - | 2 | 638 | |||
BX497188.1 | - | 2 | 106 | |||
U58914.1 | - | 2 | 152 |
Gene Symbol: CCL15-CCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027921.3 | 2 | 638 | |||
NR_027922.3 | 2 | 638 | ||||
GenBank mRNA | BC140941.1 | - | 2 | 221 | 9056753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
