siRNA Details
Gene Symbol: TMSB4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021109.3 | NP_066932.1 | 1 | 150 | ||
GenBank mRNA | AK055976.1 | - | 1 | 1224 | ||
AK057827.1 | - | 2 | 147 | |||
AK311747.1 | - | 2 | 148 | |||
AV715954.1 | - | 2 | 150 | |||
BC001631.1 | - | 1 | 131 | |||
BC016732.1 | - | 1 | 151 | |||
BC022857.1 | - | 1 | 151 | |||
BC061586.1 | - | 2 | 137 | |||
BC062436.1 | - | 1 | 151 | |||
BC064954.1 | - | 2 | 131 | |||
BC070248.1 | - | 2 | 148 | |||
BC070249.1 | - | 1 | 147 | |||
BC083509.1 | - | 1 | 147 | |||
BC092437.1 | - | 1 | 147 | |||
BC101790.1 | - | 2 | 134 | 8069296 | ||
BC101792.1 | - | 2 | 134 | 8069298 | ||
BC104464.1 | - | 1 | 101 | 40031330 | ||
BC104465.1 | - | 1 | 101 | 40031337 | ||
BC104666.1 | - | 2 | 151 | |||
BC107789.1 | - | 2 | 154 | |||
BC128102.1 | - | 2 | 119 | 40112039 | ||
BC128103.1 | - | 2 | 122 | 40112043 | ||
BC139925.1 | - | 1 | 136 | 40148985 | ||
BC141975.1 | - | 2 | 122 | 40112044 | ||
BC141976.1 | - | 2 | 122 | 40112045 | ||
BC151215.1 | - | 1 | 137 | 40148015 | ||
BT007090.1 | - | 1 | 72 | |||
M17733.1 | - | 2 | 149 | |||
X02493.1 | - | 1 | 52 |
Gene Symbol: TMSB4XP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC090885.1 | - | 11 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
