siRNA Details
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 6 | 1877 | ||
NM_001300749.1 | NP_001287678.1 | 6 | 1659 | |||
NM_139003.2 | NP_620572.1 | 5 | 1559 | |||
NM_139004.2 | NP_620573.1 | 5 | 1601 | |||
NM_139006.2 | NP_620575.1 | 6 | 1835 | |||
NM_139007.2 | NP_620576.1 | 5 | 1613 | |||
NM_139008.2 | NP_620577.1 | 5 | 1571 | |||
NM_139009.2 | NP_620578.1 | 6 | 1808 | |||
NM_139010.2 | NP_620579.1 | 4 | 1337 | |||
NM_139011.2 | NP_620580.1 | 3 | 1061 | |||
XM_011514543.2 | XP_011512845.1 | 7 | 1654 | |||
GenBank mRNA | AF144242.1 | - | 5 | 1564 | ||
BC117201.1 | - | 6 | 1752 | 40125752 | ||
BC117203.1 | - | 6 | 1752 | 40125754 | ||
U60319.1 | - | 6 | 1938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
