siRNA Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015892.1 | NP_001015892.1 | 3 | 1382 | ||
NM_003187.4 | NP_003178.1 | 3 | 1145 | |||
GenBank mRNA | BC003400.1 | - | 3 | 1073 | 3448836 | |
BC033320.1 | - | 3 | 1933 | 4837792 | ||
U21858.1 | - | 3 | 1062 | |||
U25112.1 | - | 2 | 961 | |||
U30504.1 | - | 2 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
