siRNA Details
Gene Symbol: LOC653513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003021.1 | XP_016858510.1 | 6 | 1045 | ||
XM_017003022.1 | XP_016858511.1 | 6 | 1045 | |||
XM_017003023.1 | XP_016858512.1 | 5 | 934 | |||
XM_017003024.1 | XP_016858513.1 | 6 | 1045 | |||
XM_017003025.1 | XP_016858514.1 | 6 | 1045 | |||
XM_017003026.1 | XP_016858515.1 | 6 | 1045 | |||
XM_017003027.1 | XP_016858516.1 | 6 | 880 | |||
XM_017003028.1 | XP_016858517.1 | 5 | 934 | |||
XM_017003029.1 | XP_016858518.1 | 5 | 934 | |||
XM_017003030.1 | XP_016858519.1 | 5 | 776 | |||
XM_017003031.1 | XP_016858520.1 | 6 | 880 | |||
XM_017003032.1 | XP_016858521.1 | 6 | 880 | |||
XM_017003033.1 | XP_016858522.1 | 6 | 880 | |||
XM_017003036.1 | XP_016858525.1 | 5 | 776 | |||
XM_017003037.1 | XP_016858526.1 | 5 | 776 | |||
XM_017003038.1 | XP_016858527.1 | 3 | 1655 | |||
XM_017003039.1 | XP_016858528.1 | 5 | 968 | |||
XM_017003040.1 | XP_016858529.1 | 4 | 388 | |||
GenBank mRNA | BX358804.2 | - | 1 | 79 |
Gene Symbol: LOC100996724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003064.1 | XP_016858553.1 | 6 | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
