siRNA Details
Gene Symbol: CCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002983.2 | NP_002974.1 | 3 | 598 | ||
GenBank mRNA | BC071834.1 | - | 3 | 570 | 5935287 | |
BC171831.1 | - | 3 | 532 | 9054485 | ||
BC171891.1 | - | 3 | 532 | 9054545 | ||
BX103455.1 | - | 3 | 598 | |||
D00044.1 | - | 3 | 579 | |||
M23452.1 | - | 3 | 579 | |||
M25315.1 | - | 3 | 579 | |||
X03754.1 | - | 3 | 579 |
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 3 | 584 | ||
NR_111964.1 | 3 | 776 | ||||
GenBank mRNA | BC027888.1 | - | 3 | 572 | 5222247 | |
BC107710.1 | - | 3 | 588 | 6699133 | ||
BC146914.1 | - | 3 | 535 | 9056842 | ||
X52149.1 | - | 3 | 762 |
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 3 | 574 | ||
GenBank mRNA | BC007783.2 | - | 3 | 569 | 4308472 | |
BC146919.1 | - | 3 | 535 | 9056847 | ||
BC147008.1 | - | 3 | 535 | 9056936 | ||
BC147013.1 | - | 3 | 535 | 9056941 | ||
CF128666.1 | - | 3 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
