siRNA Details
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077491.1 | NP_001070959.1 | 7 | 1275 | ||
NM_001077492.1 | NP_001070960.1 | 6 | 1207 | |||
NM_012427.4 | NP_036559.1 | 6 | 1410 | |||
XM_011526702.1 | XP_011525004.1 | 6 | 1149 | |||
XM_011526703.2 | XP_011525005.1 | 7 | 1229 | |||
GenBank mRNA | AF168768.2 | - | 6 | 1226 | ||
AK223421.1 | - | 6 | 1358 | |||
AY279380.1 | - | 6 | 1184 | |||
AY279381.1 | - | 7 | 1252 | |||
AY359010.1 | - | 6 | 1410 | |||
AY461805.1 | - | 4 | 1153 | |||
BC008036.2 | - | 6 | 1341 | 2989806 |
