siRNA Details
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270621.1 | NP_001257550.1 | 11 | 2101 | ||
NM_031075.2 | NP_112337.2 | 12 | 2173 | |||
GenBank mRNA | X90651.1 | - | 12 | 2173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
