siRNA Details
Gene Symbol: ATP2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199179.1 | NP_001186108.1 | 28 | 3601 | ||
NM_001199181.1 | NP_001186110.1 | 27 | 3590 | |||
NM_001199184.1 | NP_001186113.1 | 27 | 3463 | |||
NM_014382.3 | NP_055197.2 | 27 | 3514 | |||
GenBank mRNA | AB037768.1 | - | 28 | 3602 | ||
AB209265.1 | - | 16 | 3362 | |||
AF181120.1 | - | 27 | 3427 | |||
AF189723.2 | - | 27 | 3345 | |||
AJ010953.1 | - | 13 | 1924 | |||
AK001684.1 | - | 15 | 2122 | |||
AK304175.1 | - | 27 | 3319 | |||
BU188061.1 | - | 1 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
