siRNA Details
Gene Symbol: NR6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278546.1 | NP_001265475.1 | 10 | 3118 | ||
NM_001489.4 | NP_001480.3 | 10 | 3115 | |||
NM_033334.3 | NP_201591.2 | 10 | 3130 | |||
XM_005251917.4 | XP_005251974.1 | 10 | 3076 | |||
XM_005251918.4 | XP_005251975.1 | 9 | 4299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
