siRNA Details
Gene Symbol: OR7E91P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002185.3 | 3 | 816 | |||
GenBank mRNA | BC014374.1 | - | 3 | 819 | ||
BC030991.1 | - | 3 | 855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
