siRNA Details
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 19 | 2271 | ||
NM_001321081.1 | NP_001308010.1 | 20 | 2335 | |||
NM_001321082.1 | NP_001308011.1 | 19 | 2242 | |||
NM_001991.4 | NP_001982.2 | 20 | 2362 | |||
XM_005257145.2 | XP_005257202.1 | 18 | 2117 | |||
XM_011524517.2 | XP_011522819.1 | 19 | 2210 | |||
XM_017024350.1 | XP_016879839.1 | 19 | 2189 | |||
XM_017024351.1 | XP_016879840.1 | 18 | 2096 | |||
XM_017024352.1 | XP_016879841.1 | 17 | 1947 | |||
GenBank mRNA | AB002386.1 | - | 19 | 2274 | ||
AK295626.1 | - | 19 | 2224 | |||
AK295853.1 | - | 18 | 2062 | |||
AK299887.1 | - | 19 | 2177 | |||
AK304835.1 | - | 19 | 2175 | |||
AL833159.1 | - | 20 | 2282 | |||
BC015882.1 | - | 20 | 2296 | 3908776 | ||
BT009782.1 | - | 18 | 2174 | |||
U50315.1 | - | 19 | 2280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
