siRNA Details
Gene Symbol: UBE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003337.3 | NP_003328.1 | 4 | 627 | ||
GenBank mRNA | AK312012.1 | - | 4 | 291 | ||
BC005979.1 | - | 4 | 234 | 4105643 | ||
BC008404.1 | - | 4 | 294 | 4278820 | ||
BC008470.1 | - | 4 | 294 | 4281004 | ||
BT007071.1 | - | 4 | 207 | |||
CR407634.1 | - | 4 | 207 | |||
M74525.1 | - | 4 | 628 | |||
X53251.1 | - | 4 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
