siRNA Details
Gene Symbol: UBE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003337.3 | NP_003328.1 | 6 | 1793 | ||
GenBank mRNA | BU954901.1 | - | 1 | 141 | ||
M74525.1 | - | 6 | 1793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
