siRNA Details
Gene Symbol: LOC105376114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135839.1 | 2 | 513 | |||
NR_135840.1 | 2 | 158 | ||||
GenBank mRNA | AK300656.1 | - | 2 | 150 | ||
HY009529.1 | - | 2 | 514 |
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243723.1 | NP_001230652.1 | 4 | 705 | ||
NM_001320676.1 | NP_001307605.1 | 3 | 373 | |||
NM_001320677.1 | NP_001307606.1 | 4 | 685 | |||
NM_001320678.1 | NP_001307607.1 | 2 | 258 | |||
NM_006515.3 | NP_006506.3 | 3 | 1122 | |||
XM_006713292.3 | XP_006713355.1 | 3 | 458 | |||
XM_006713294.3 | XP_006713357.1 | 4 | 747 | |||
XM_017007032.1 | XP_016862521.1 | 3 | 1185 | |||
GenBank mRNA | AF054989.1 | - | 3 | 1048 | ||
AK122967.1 | - | 1 | 1318 | |||
AK222734.1 | - | 3 | 1058 | |||
AK297865.1 | - | 4 | 620 | |||
AK302296.1 | - | 4 | 639 | |||
AK307537.1 | - | 3 | 1122 | |||
AK311418.1 | - | 3 | 1060 | |||
AY952295.1 | - | 3 | 1027 | |||
BC008931.2 | - | 3 | 292 | |||
U80776.1 | - | 3 | 1039 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
