siRNA Details
Gene Symbol: UBR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282873.1 | NP_001269802.1 | 41 | 6206 | ||
NM_015902.5 | NP_056986.2 | 41 | 6206 | |||
XM_005250962.4 | XP_005251019.1 | 20 | 2967 | |||
XM_011517104.2 | XP_011515406.2 | 40 | 6191 | |||
XM_011517105.2 | XP_011515407.2 | 40 | 6191 | |||
XM_011517106.2 | XP_011515408.2 | 40 | 6173 | |||
XM_017013533.1 | XP_016869022.1 | 40 | 6191 | |||
XM_017013534.1 | XP_016869023.1 | 40 | 6191 | |||
GenBank mRNA | AB020703.3 | - | 41 | 6206 | ||
AF006010.2 | - | 41 | 5784 | |||
BC137234.1 | - | 41 | 5778 | 9021231 | ||
U95000.1 | - | 41 | 5748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
