siRNA Details
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320687.1 | NP_001307616.1 | 12 | 2138 | ||
NM_016166.2 | NP_057250.1 | 11 | 1539 | |||
XM_011522126.2 | XP_011520428.1 | 13 | 2246 | |||
XM_011522127.2 | XP_011520429.1 | 11 | 2023 | |||
XM_017022688.1 | XP_016878177.1 | 11 | 2017 | |||
XM_017022689.1 | XP_016878178.1 | 11 | 2030 | |||
XM_017022690.1 | XP_016878179.1 | 11 | 1835 | |||
XM_017022691.1 | XP_016878180.1 | 11 | 1609 | |||
GenBank mRNA | AF077951.1 | - | 11 | 1388 | ||
AF167160.1 | - | 11 | 1487 | |||
AK094641.1 | - | 12 | 2138 | |||
AK314515.1 | - | 11 | 1414 | |||
AY826819.1 | - | 11 | 1388 | |||
BC118587.1 | - | 11 | 1441 | 40085010 | ||
BC121797.1 | - | 11 | 1438 | 40085007 | ||
EF487997.1 | - | 11 | 1391 | |||
EF487998.1 | - | 11 | 1391 | |||
U78524.1 | - | 10 | 1373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
