siRNA Details
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320687.1 | NP_001307616.1 | 7 | 1500 | ||
NM_016166.2 | NP_057250.1 | 6 | 901 | |||
XM_011522126.2 | XP_011520428.1 | 8 | 1608 | |||
XM_011522127.2 | XP_011520429.1 | 6 | 1385 | |||
XM_017022688.1 | XP_016878177.1 | 6 | 1379 | |||
XM_017022689.1 | XP_016878178.1 | 6 | 1392 | |||
XM_017022690.1 | XP_016878179.1 | 6 | 1197 | |||
XM_017022691.1 | XP_016878180.1 | 6 | 971 | |||
GenBank mRNA | AF077951.1 | - | 6 | 753 | ||
AF167160.1 | - | 6 | 849 | |||
AK094641.1 | - | 7 | 1500 | |||
AK314515.1 | - | 6 | 776 | |||
AY826819.1 | - | 6 | 750 | |||
BC053685.1 | - | 6 | 754 | |||
BC118587.1 | - | 6 | 803 | 40085010 | ||
BC121797.1 | - | 6 | 800 | 40085007 | ||
EF487997.1 | - | 6 | 753 | |||
EF487998.1 | - | 6 | 753 | |||
FJ997900.1 | - | 6 | 753 | |||
U78524.1 | - | 5 | 735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
