siRNA Details
Gene Symbol: ASH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018489.2 | NP_060959.2 | 5 | 6065 | ||
XM_006711450.3 | XP_006711513.1 | 5 | 5756 | |||
XM_006711451.3 | XP_006711514.1 | 6 | 5843 | |||
XM_006711452.3 | XP_006711515.1 | 5 | 6065 | |||
XM_011509770.2 | XP_011508072.1 | 4 | 1436 | |||
XM_017001784.1 | XP_016857273.1 | 5 | 5842 | |||
XM_017001785.1 | XP_016857274.1 | 5 | 5772 | |||
XM_017001786.1 | XP_016857275.1 | 5 | 6065 | |||
XM_017001787.1 | XP_016857276.1 | 5 | 6065 | |||
XM_017001788.1 | XP_016857277.1 | 2 | 560 | |||
GenBank mRNA | AF257305.1 | - | 5 | 5894 | ||
CN365042.1 | - | 1 | 69 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
