siRNA Details
Gene Symbol: USP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525067.2 | XP_011523369.1 | 21 | 3746 | ||
XM_011525071.2 | XP_011523373.1 | 16 | 2567 | |||
XM_017024900.1 | XP_016880389.1 | 21 | 3870 | |||
GenBank mRNA | AB040886.1 | - | 21 | 5840 | ||
AK022913.1 | - | 22 | 3640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
